missing translation for 'onlineSavingsMsg'
Learn More

DSE Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1696.32 NOK - 4796.50 NOK

Specifications

Antigen DSE
Concentration 0.13 mg/mL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16806853
View Documents
Proteintech
10452-1-AP-20UL
20 μL
1696.32 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16896843
View Documents
Proteintech
10452-1-AP-150UL
150 μL
4796.50 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Converts D-glucuronic acid to L-iduronic acid (IdoUA) residues.
Specifications

Specifications

DSE
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
Q9UL01
29940
DSE Fusion Protein Ag0695
Primary
-20°C
DSE
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
dermatan sulfate epimerase, DS epimerase, DSE, DSEPI, SART 2, SART2
DSE
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.