Learn More
LABPLAS Dry Polyurethane Sani-Stick
Biocide free dry sponge with handle for sampling surfaces.
2890.00 NOK
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
17900205
|
LABPLAS
KSS-67910 |
None |
2890.00 NOK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17990195
|
LABPLAS
KSS-67915 |
Gloves |
2890.00 NOK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Sani-Stick
- Dry polyurethane sponge with handle
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Specifications
Hygiene Sponge Sampling Kit | |
Sterile |
Product Title
LABPLAS Dry Polyurethane Sani-Stick