Learn More
LABPLAS Dry Cellulose Sani-Sponge
Cellulose sponge for sampling surfaces.
Brand: LABPLAS KSS-61105
This item is not returnable.
View return policy
Description
- SANI-SPONGE
- Dry sponge
- Dimensions: 114 x 229 mm
- Holds: 450 mL
Specifications
Hygiene Sponge Sampling Kit | |
Sterile |