Learn More
Thermo Scientific™ Drums for Digital Cel-Gro Tissue Culture Rotators
Accessory for use with Digital Cel-Gro Tissue Culture Rotator, 120V and Digital Cel-Gro Tissue Culture Rotator, 230V
2003.29 NOK - 3902.97 NOK
Product Code | Brand | Description | Diameter (Metric) | For Use With (Application) | No. of Positions | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Diameter (Metric) | For Use With (Application) | No. of Positions | Price | Quantity & Availability | ||
17378272
|
Thermo Scientific™
88882149 |
50 x 1.5/2.0 mL rotator drum | 290 mm | 1.5, 2.0 mL tubes | 50 |
2498.36 NOK
Each |
In Stock
Log in to see stock. |
||
17388272
|
Thermo Scientific™
88882150 |
30 x 15 mL rotator drum | 290 mm | 15 mL tubes | 30 |
2003.29 NOK
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||
17398272
|
Thermo Scientific™
88882151 |
15 x 50 mL rotator drum | 290 mm | 50 mL tubes | 15 |
2003.29 NOK
Each |
Estimated Shipment: 02-12-2024
Log in to see stock. |
||
17308282
|
Thermo Scientific™
88882152 |
10 x 10/15 mL and 20 x 5/7 mL rotator drum | 290 mm | 10/15 mL, 5/7 mL tubes | 30 |
2003.29 NOK
Each |
Estimated Shipment: 23-10-2024
Log in to see stock. |
||
17318282
|
Thermo Scientific™
88882153 |
64 x diameter 14 mm tube rotator drum | 310 mm | diameter 14 mm tube | 64 |
2003.29 NOK
Each |
In Stock
Log in to see stock. |
||
17328282
|
Thermo Scientific™
88882154 |
64 x diameter 18.5 mm tube rotator drum | 310 mm | diameter 18.5 mm tube | 64 |
2360.20 NOK
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
||
17338282
|
Thermo Scientific™
88882155 |
142 x diameter 17 mm tube rotator drum | 380 mm | diameter 17 mm tube | 142 |
3902.97 NOK
Each |
Estimated Shipment: 01-11-2024
Log in to see stock. |
||
17348282
|
Thermo Scientific™
88882156 |
76 x diameter 26 mm tube rotator drum | 380 mm | diameter 26 mm tube | 76 |
3430.93 NOK
Each |
Estimated Shipment: 23-10-2024
Log in to see stock. |
||
17358282
|
Thermo Scientific™
88882157 |
60 x 30 diameter mm tube rotator drum | 380 mm | diameter 30 mm tube | 60 |
3430.93 NOK
Each |
Estimated Shipment: 23-10-2024
Log in to see stock. |
||
Description
Accessory for use with Digital Cel-Gro Tissue Culture Rotator, 120V and Digital Cel-Gro Tissue Culture Rotator, 230V
Specifications
Rotating Drum |