Learn More
Multiroir Controlec™ Drawer Separation Inserts for Crystal Multiroir 10000
Creates a compartment within a 10L drawer. Multiroir Controlec™ Drawer Separation Inserts for Crystal Multiroir 10000 are a collection of longitudinal and cross pieces to create customized sectioned storage. Each 100mm tall piece is made of 7mm-thick, crystal clear polystyrene.
Brand:  Multiroir Controlec™ 11153
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11856860
  • 217.00 NOK / Pack of 10
Estimated Shipment: 08-08-2024
Description
Crystal Multiroir are high quality transparent drawer cabinets. They are completely modular and can be combined at will from all sides to save space. Each drawer can be customized with removable partitions to create any size of compartment for partitioned storage.
  • Can be placed on cabinets, shelves or the floor
  • Each unit includes a handle with label holder
  • High transparency Crystal Polystyrene material
  • Shock resistant
  • UV resistant
  • Soap washable
  • Fall arrest stops on each drawer
  • Easy to assemble, tool-free, slide from right to left and from bottom to top
Specifications
Specifications
10/Pack
1/3 Transverse Drawer Separation Insert
7 mm
Transparent
80 mm
Polystyrene
Sectioned storage
100 mm
Crystal Mutiroir 10000 (10 L) Drawer, Transverse