Learn More
Dräger™ CMS Chip Measurement System
Uses an analyzer and substance-specific chips to obtain accurate, reliable measurements of gas and vapor concentrations
Brand:  Dräger™ 6405300
 View more versions of this product
Product Code. 10629932
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes: Comes with carrying case and batteries. Optional remote system (sold separately) includes an additional pump, a screwdriver, and a 9 ft. (3m) extension hose to sample contaminants in inaccessible areas.
Requires : CMS chips (sold separately)
Description
Description
Bar code on each chip contains specific information such as gas type, measurement range, and other necessary parameters for measurement evaluation; when a chip is inserted into the analyzer, the bar code is read and interpreted. Choose from more than 50 interchangeable chips (sold separately).
  • Mass-flow-regulated pump system draws a constant flow of sample through the chip, compensating for any pressure fluctuation
  • Easy to use, with simple menu commands and one-switch operation
  • Chip contains 10 measurement channels filled with a substance-specific reagent that provides a quick reaction with the contaminant
  • Intelligent interaction ensures high level of accuracy
  • Data recorder gives contaminant, concentration, date, time, and location; stores up to 50 measurements
Order Info
Compact, rugged housing measures approximately 1.5L x 3.5W x 8 in. H (3.8 x 8.9 x 20.3cm). Net weight: 1.63 lb. (730g). Uses four AA alkaline batteries (included).
Certifications
Intrinsically safe and certified according to UL, MSHA and CENELEC standards. (UL and CSA Classified as Intrinsically Safe for use in Class I, Div. 1, Groups A-D, T4.)
Specifications
Specifications
CMS Analyzer
Documents
One moment while we fetch your results.
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Dräger™ CMS Chip Measurement System > Analyzer