Learn More
Bochem™ Double-Sleeve Malleable Cast Iron Bosshead
Manufactured from malleable cast iron. BOCHEM™ Double-Sleeve Malleable Cast Iron Bosshead is DIN 12895 certified.
Brand:  Bochem™ 5359
Product Code. 10652593
  • 164.00 NOK / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Description
  • Thumb screw M8
  • Malleable cast iron (DIN 12895)
  • Chromium plated
Specifications
Specifications
1 Ea.
Malleable Cast Iron
20 mm
Double Sleeve
20 mm