missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Double Quint Blood Bag Adapter

Double Quint Blood Bag Adapter for XXL size, HD (110 x 88 mm).

Brand:  Thermo Scientific 75003899

Product Code. 17868800

  • 3421.36 NOK / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Double Quint Blood Bag Adapter for XXL size, HD (110 x 88 mm)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blood Bag Adapter
Plastic
12 or 16 x 500 mL
Not Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Double Quint Blood Bag Adapter > Double Quint Blood Bag Adapter; Capacity: 12 or 16 x 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.