Learn More
SI Analytics™ Double-Platinum Single Electrode
Especially useful for the separation of combination electrodes into separate measuring and reference electrodes. SI Analytics™ Double-Platinum Single Electrode is for special applications and makes complex applications even more reliable.
Brand: SI Analytics™ 285103512
This item is not returnable.
View return policy
Description
- The combination of pure measuring electrodes for i.e. ion-selective measurements or special constructions with one reference electrode
- Redox titrations with a constant pH value with glass electrodes as reference and redox single sensors as measuring electrode
- Application of double-electrolyte electrodes for an optimal adaptation of reference electrolyte to the measuring media.
Specifications
Plug Head | |
Single Electrode | |
-30°C to +135°C | |
12 mm |
Glass | |
Pt 1200 | |
120 mm |
Product Content Correction
Product Title
SI Analytics™ Double-Platinum Single Electrode > LengthMetric: 120mm