Learn More
DLK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10636-1-AP-20UL
 View more versions of this product
Product Code. 16897133
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
This gene encodes a transmembrane protein containing six epidermal growth factor repeats. The protein is involved in the differentiation of several cell types, including adipocytes; it is also thought to be a tumor suppressor. It is one of several imprinted genes located in a region of on chr 14q32. Certain mutations in this imprinted region can cause phenotypes similar to maternal and paternal uniparental disomy of chromosome 14 (UPD14). This gene is expressed from the paternal allele. A polymorphism within this gene has been associated with child and adolescent obesity. The mode of inheritance for this polymorphism is polar overdomice; this non-Mendelian inheritance pattern was first described in sheep with the callipyge phenotype, which is characterized by muscle hypertrophy and decreased fat mass.
Specifications
Specifications
DLK1
Polyclonal
Unconjugated
DLK1
DLK, DLK 1, DLK1, FA1, pG2, Pref 1, PREF1, Protein delta homolog 1, ZOG
Rabbit
Antigen Affinity Chromatography
RUO
114587, 13386, 8788
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.34 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P80370, Q09163
DLK1
DLK1 Fusion Protein Ag0991
20 μL
Primary
Human, Mouse, Rat