missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10636-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a transmembrane protein containing six epidermal growth factor repeats. The protein is involved in the differentiation of several cell types, including adipocytes; it is also thought to be a tumor suppressor. It is one of several imprinted genes located in a region of on chr 14q32. Certain mutations in this imprinted region can cause phenotypes similar to maternal and paternal uniparental disomy of chromosome 14 (UPD14). This gene is expressed from the paternal allele. A polymorphism within this gene has been associated with child and adolescent obesity. The mode of inheritance for this polymorphism is polar overdomice; this non-Mendelian inheritance pattern was first described in sheep with the callipyge phenotype, which is characterized by muscle hypertrophy and decreased fat mass.Specifications
DLK1 | |
Polyclonal | |
Unconjugated | |
DLK1 | |
DLK, DLK 1, DLK1, FA1, pG2, Pref 1, PREF1, Protein delta homolog 1, ZOG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114587, 13386, 8788 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P80370, Q09163 | |
DLK1 | |
DLK1 Fusion Protein Ag0991 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction