Learn More
DLin-KC2-DMA, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
DLin-KC2-DMA is a cationic/ionizable lipid for siRNA delivery.
Specifications
Specifications
Chemical Name or Material | DLin-KC2-DMA |
CAS | 1190197-97-7 |
Color | Colorless |
Purity Grade Notes | Research |
Molecular Formula | C43H79NO2 |
Quantity | 50 mg |
Solubility Information | Ethanol : 100 mg/mL (155.74 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (155.74 mM; Need ultrasonic) |
SMILES | CCCCC/C=C\C/C=C\CCCCCCCCC1(CCCCCCCC/C=C\C/C=C\CCCCC)OC(CCN(C)C)CO1 |
Molecular Weight (g/mol) | 642.09 |
Formula Weight | 642.09 |
Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
DLin-KC2-DMA, MedChemExpress