Learn More
Thermo Scientific Chemicals DL-Tyrosine, 99%
785.20 NOK - 2129.94 NOK
Chemical Identifiers
CAS | 556-03-6 |
---|---|
Molecular Formula | C9H11NO3 |
Molecular Weight (g/mol) | 181.19 |
MDL Number | MFCD00063074 |
InChI Key | OUYCCCASQSFEME-UHFFFAOYNA-N |
Synonym | dl-tyrosine, h-dl-tyr-oh, 2-amino-3-4-hydroxyphenyl propanoic acid, tyrosin, tyrosine, dl, l-tyrosine, free base, tirosina, l-tryosine, 3-4-hydroxyphenyl-dl-alanine, benzenepropanoic acid, s |
PubChem CID | 1153 |
ChEBI | CHEBI:18186 |
IUPAC Name | 2-amino-3-(4-hydroxyphenyl)propanoic acid |
SMILES | NC(CC1=CC=C(O)C=C1)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10675792
|
Thermo Scientific Chemicals
163570250 |
25 g | Glass Bottle |
785.20 NOK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10245640
|
Thermo Scientific Chemicals
163571000 |
100 g | Glass Bottle |
2129.94 NOK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
556-03-6 | |
181.19 | |
OUYCCCASQSFEME-UHFFFAOYNA-N | |
1153 | |
2-amino-3-(4-hydroxyphenyl)propanoic acid |
C9H11NO3 | |
MFCD00063074 | |
dl-tyrosine, h-dl-tyr-oh, 2-amino-3-4-hydroxyphenyl propanoic acid, tyrosin, tyrosine, dl, l-tyrosine, free base, tirosina, l-tryosine, 3-4-hydroxyphenyl-dl-alanine, benzenepropanoic acid, s | |
CHEBI:18186 | |
NC(CC1=CC=C(O)C=C1)C(O)=O |
Specifications
556-03-6 | |
99% | |
Authentic | |
MFCD00063074 | |
14, 9839 | |
OUYCCCASQSFEME-UHFFFAOYNA-N | |
2-amino-3-(4-hydroxyphenyl)propanoic acid | |
1153 | |
181.19 | |
Powder |
White | |
C9H11NO3 | |
4-(HO)C6H4CH2CH(NH2)CO2H | |
14, 621 | |
dl-tyrosine, h-dl-tyr-oh, 2-amino-3-4-hydroxyphenyl propanoic acid, tyrosin, tyrosine, dl, l-tyrosine, free base, tirosina, l-tryosine, 3-4-hydroxyphenyl-dl-alanine, benzenepropanoic acid, s | |
NC(CC1=CC=C(O)C=C1)C(O)=O | |
181.19 | |
CHEBI:18186 | |
98.5 to 101.5% | |
DL-Tyrosine, 99% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 209-113-1
missing translation for 'tsca' : TSCA
- DL-Tyrosine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only