missing translation for 'onlineSavingsMsg'
Learn More

DL-Thioctic acid, 98-99%, MP Biomedicals™

Product Code. 11388739 Shop All MP Biomedicals Products
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1077-28-7
Molecular Formula C8H14O2S2
Molecular Weight (g/mol) 206.32
MDL Number MFCD00005474
InChI Key AGBQKNBQESQNJD-UHFFFAOYNA-N
Synonym dl-thioctic acid, thioctic acid, alpha-lipoic acid, 5-1,2-dithiolan-3-yl pentanoic acid, dl-alpha-lipoic acid, 1,2-dithiolane-3-pentanoic acid, 6,8-thioctic acid, thioctacid, 6-thioctic acid, alpha lipoic acid
PubChem CID 864
ChEBI CHEBI:16494
IUPAC Name 5-(1,2-dithiolan-3-yl)pentanoic acid
SMILES OC(=O)CCCCC1CCSS1

Specifications

Color Yellow
Assay Percent Range 98 to 99%
Quantity 5 g
Formula Weight 206.3
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Chemical Name or Material DL-Thioctic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DL-Thioctic acid, 98-99%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.