missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Serine, ≥98%, MP Biomedicals™
583.00 NOK - 6513.70 NOK
Chemical Identifiers
CAS | 302-84-1 |
---|---|
Molecular Formula | C3H7NO3 |
Molecular Weight (g/mol) | 105.093 |
InChI Key | MTCFGRXMJLQNBG-UHFFFAOYSA-N |
Synonym | dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid |
PubChem CID | 617 |
ChEBI | CHEBI:17822 |
IUPAC Name | 2-amino-3-hydroxypropanoic acid |
SMILES | C(C(C(=O)O)N)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11466242
|
MP Biomedicals
0210286825 |
25 g |
583.00 NOK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11476242
|
MP Biomedicals
0210286880 |
100 g |
927.50 NOK
100g |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||||
11486242
|
MP Biomedicals
0210286891 |
1 kg |
6513.70 NOK
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
302-84-1 | |
105.093 | |
dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid | |
CHEBI:17822 | |
C(C(C(=O)O)N)O |
C3H7NO3 | |
MTCFGRXMJLQNBG-UHFFFAOYSA-N | |
617 | |
2-amino-3-hydroxypropanoic acid |
Specifications
302-84-1 | |
≥98% | |
dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid | |
C(C(C(=O)O)N)O | |
105.093 | |
CHEBI:17822 | |
1.537g/mL | |
DL-Serine |
White | |
C3H7NO3 | |
MTCFGRXMJLQNBG-UHFFFAOYSA-N | |
2-amino-3-hydroxypropanoic acid | |
617 | |
105.1 | |
Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 206-130-6
Recommended Storage : Store at room temperature.