missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Dithiothreitol, 99.5%, MP Biomedicals™
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
Specifications
Specifications
Color | White |
Assay Percent Range | >99% |
Quantity | 50 g |
Formula Weight | 154.2 |
Grade | Molecular Biology Reagent |
Physical Form | Powder |
Chemical Name or Material | DL-Dithiothreitol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'