Learn More
DL-Cystine, Thermo Scientific Chemicals
639.00 NOK - 6021.00 NOK
Chemical Identifiers
CAS | 923-32-0 |
---|---|
Molecular Formula | C6H12N2O4S2 |
Molecular Weight (g/mol) | 240.292 |
MDL Number | MFCD00084652 |
InChI Key | LEVWYRKDKASIDU-UHFFFAOYSA-N |
Synonym | dl-cystine, cystine, d-cystine, 3,3'-disulfanediylbis 2-aminopropanoic acid, dicysteine, cystin, h-dl-cys-oh 2, .beta.,.beta.'-dithiodialanine, cystine, dl, cystine, l |
PubChem CID | 595 |
ChEBI | CHEBI:17376 |
IUPAC Name | 2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid |
SMILES | C(C(C(=O)O)N)SSCC(C(=O)O)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15485759
|
Thermo Scientific Alfa Aesar
J63564.06 |
5 g |
639.00 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15495759
|
Thermo Scientific Alfa Aesar
J63564.14 |
25 g |
1809.00 NOK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15405769
|
Thermo Scientific Alfa Aesar
J63564.22 |
100 g |
6021.00 NOK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DL-cystine is used in the preparation of sulfur-containing dimeric and monomeric surfactants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-cystine is used in the preparation of sulfur-containing dimeric and monomeric surfactants.
Solubility
Soluble in dilute acid and alkali solution, highly soluble in water, not soluble in alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
923-32-0 | |
240.292 | |
LEVWYRKDKASIDU-UHFFFAOYSA-N | |
595 | |
2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid |
C6H12N2O4S2 | |
MFCD00084652 | |
dl-cystine, cystine, d-cystine, 3,3'-disulfanediylbis 2-aminopropanoic acid, dicysteine, cystin, h-dl-cys-oh 2, .beta.,.beta.'-dithiodialanine, cystine, dl, cystine, l | |
CHEBI:17376 | |
C(C(C(=O)O)N)SSCC(C(=O)O)N |
Specifications
227°C (decomposition) | |
White | |
MFCD00084652 | |
14,2782 | |
Soluble in dilute acid and alkali solution,highly soluble in water,not soluble in alcohol. | |
C(C(C(=O)O)N)SSCC(C(=O)O)N | |
240.292 | |
CHEBI:17376 | |
Powder |
923-32-0 | |
C6H12N2O4S2 | |
1728095 | |
dl-cystine, cystine, d-cystine, 3,3'-disulfanediylbis 2-aminopropanoic acid, dicysteine, cystin, h-dl-cys-oh 2, .beta.,.beta.'-dithiodialanine, cystine, dl, cystine, l | |
LEVWYRKDKASIDU-UHFFFAOYSA-N | |
2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid | |
595 | |
240.3 | |
DL-Cystine |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 213-094-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only