missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-beta-Homoproline hydrochloride, 97% Product Code.: 15474367

Thermo Scientific Chemicals DL-beta-Homoproline hydrochloride, 97%

Product Code. 15474367
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15474367

Brand: Thermo Scientific Chemicals H31749.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71985-79-0
Molecular Formula C6H12ClNO2
Molecular Weight (g/mol) 165.617
MDL Number MFCD11506301
InChI Key VQDACVOAOJQTPR-UHFFFAOYSA-N
Synonym pyrrolidin-2-yl-acetic acid hydrochloride, 2-pyrrolidin-2-yl acetic acid hydrochloride, pyrrolidin-2-ylacetic acid hydrochloride, 2-pyrrolidineacetic acid hydrochloride, 2-pyrrolidin-2-yl acetic acid hcl, h-d-beta-hopro-oh.hcl, 2-pyrrolidineacetic acid hcl, dl-beta-homoproline hydrochloride, 2-pyrrolidineacetic acid, hydrochloride, 2-pyrrolidinylacetic acid hydrochlorideShow MoreShow Less
PubChem CID 12514707
IUPAC Name 2-pyrrolidin-2-ylacetic acid;hydrochloride
SMILES C1CC(NC1)CC(=O)O.Cl

Specifications

Melting Point 171°C to 174°C
Quantity 1 g
Formula Weight 165.62
Percent Purity 97%
Chemical Name or Material DL-beta-Homoproline hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10183353 Certificate TypeCertificate of Analysis Date22/11/2023 Catalog NumberH31749.MD,H31749.03
Lot Number10183284 Certificate TypeCertificate of Analysis Date22/11/2023 Catalog NumberH31749.MD,H31749.03
Lot Number10183353 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH31749.MD,H31749.03
Lot Number10183284 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH31749.MD,H31749.03
Lot Number10160386 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH31749.MD,H31749.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-beta-Homoproline hydrochloride, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.