Learn More
DL-Alanine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals
CAS: 617-27-6 | C5H12ClNO2 | 153.61 g/mol
689.00 NOK - 2785.00 NOK
Chemical Identifiers
CAS | 617-27-6 |
---|---|
Molecular Formula | C5H12ClNO2 |
Molecular Weight (g/mol) | 153.61 |
MDL Number | MFCD00013018 |
InChI Key | JCXLZWMDXJFOOI-UHFFFAOYNA-N |
Synonym | ethyl 2-aminopropanoate hydrochloride, dl-alanine ethyl ester hydrochloride, h-dl-ala-oet.hcl, h-dl-ala-oet hydrochloride, d,l-alanine ethyl ester hydrochloride, h-dl-ala-oet hcl, l-alanine ethyl ester hcl, alanine, ethyl ester, hydrochloride 1:1, alanine, ethyl ester, hydrochloride, ethyl alaninate hydrochloride |
PubChem CID | 102560 |
IUPAC Name | ethyl 2-aminopropanoate;hydrochloride |
SMILES | Cl.CCOC(=O)C(C)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11333146
|
Thermo Scientific Alfa Aesar
L11020.06 |
5 g |
689.00 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343146
|
Thermo Scientific Alfa Aesar
L11020.14 |
25 g |
2785.00 NOK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
617-27-6 | |
153.61 | |
JCXLZWMDXJFOOI-UHFFFAOYNA-N | |
102560 | |
Cl.CCOC(=O)C(C)N |
C5H12ClNO2 | |
MFCD00013018 | |
ethyl 2-aminopropanoate hydrochloride, dl-alanine ethyl ester hydrochloride, h-dl-ala-oet.hcl, h-dl-ala-oet hydrochloride, d,l-alanine ethyl ester hydrochloride, h-dl-ala-oet hcl, l-alanine ethyl ester hcl, alanine, ethyl ester, hydrochloride 1:1, alanine, ethyl ester, hydrochloride, ethyl alaninate hydrochloride | |
ethyl 2-aminopropanoate;hydrochloride |
Specifications
84°C to 86°C | |
C5H12ClNO2 | |
3654425 | |
JCXLZWMDXJFOOI-UHFFFAOYNA-N | |
ethyl 2-aminopropanoate;hydrochloride | |
102560 | |
Hygroscopic | |
DL-Alanine ethyl ester hydrochloride |
617-27-6 | |
MFCD00013018 | |
ethyl 2-aminopropanoate hydrochloride, dl-alanine ethyl ester hydrochloride, h-dl-ala-oet.hcl, h-dl-ala-oet hydrochloride, d,l-alanine ethyl ester hydrochloride, h-dl-ala-oet hcl, l-alanine ethyl ester hcl, alanine, ethyl ester, hydrochloride 1:1, alanine, ethyl ester, hydrochloride, ethyl alaninate hydrochloride | |
Cl.CCOC(=O)C(C)N | |
153.61 | |
153.61 | |
99% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 210-507-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- DL-Alanine ethyl ester hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : DL-Alanine ethyl ester hydrochloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.