missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ DJ462O23.2 Recombinant Protein

Brand:  Abnova™ H00057185-P01.25ug

 View more versions of this product

Product Code. 16122593

  • 6245.00 NOK / 25µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH01265
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
48.4
8
Glutathione Sepharose 4 Fast Flow
25 ug
SAVSEASFSYKENLIGALLAIFGHLVVSIALNLQKYCHIRLAGSKDPRAYFKTKTWWLGLFLMLLGELGVFASYAFAPLSLIVPLSAVSVIASAIIGIIFIKEKWKPKDFLRRYVLSFVGCGLAVVGTYLLVTFAPNSHEKMTGENVTRHLVSWPFLLYMLVEIILFCLLLYFYKEKNANNIVVILLLVALLVLLCHPGWSAVVQS
DJ462O23.2/DKFZp686E22155
NPAL3
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
57185
DJ462O23.2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NPAL3
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers