Learn More
Lenz Laborglasintrumente Distilling Links with Liebig Condenser and GL Thread
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
1215.00 NOK - 1320.00 NOK
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
11562324
|
Lenz Laborglasintrumente
05050414 |
29/32 NS |
1320.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11552324
|
Lenz Laborglasintrumente
05050311 |
14/23 NS |
1215.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Distilling links with Liebig condenser
- Outlet with NS cone, water connection with thread GL14
- With socket NS 14/23 for distillation thermometers of 55 mm built-in length
- Made of DURAN™ tubing
Specifications
Duran Glass | |
1 Ea. |
Product Title
Lenz Laborglasintrumente Distilling Links with Liebig Condenser and GL Thread