Learn More
Dissolved Oxygen Electrolyte Solution, Reagecon™ Product Code.: 11583172
Dissolved Oxygen Electrolyte Solution, Reagecon™
BIOPHARMA
Product Code. 11583172 Shop All Reagecon Products
100 mL
Click to view available options
Quantity:
100 mL
Product Code. 11583172
Brand: Reagecon EFSDO
Specifications
Quantity 100 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberEFSDO23K1 Certificate TypeCertificate of Analysis Date11/12/2024 Catalog NumberEFSDO
Lot NumberEFSDO24E1 Certificate TypeCertificate of Analysis Date08/10/2024 Catalog NumberEFSDO
Lot NumberEFSDO22C1 Certificate TypeCertificate of Analysis Date09/03/2022 Catalog NumberEFSDO
Lot NumberEFSDO20H1 Certificate TypeCertificate of Analysis Date20/10/2021 Catalog NumberEFSDO
Product Content Correction
Product Title
Dissolved Oxygen Electrolyte Solution, Reagecon™ >