Learn More
Dissecting Set in Plastic Wallet
Use this dissecting kit for a variety of applications. Dissecting Sets in Plastic Wallets are suitable for theatre, ward, veterinary, dental and chiropody use.
Brand: S Murray SAMCO Y261/71
This item is not returnable.
View return policy
Description
Basic, student quality stainless steel instruments in wallet
Set includes:
- 1 forceps (125mm, straight, fine point)
- 1 scalpel (blade length 38mm)
- 1 forceps (125mm, straight, blunt tip)
- 1 scissors (125mm straight, fine point)
- 2 needles in metal handle
Specifications
Plastic | |
Stainless Steel | |
Dissecting Set Wallet |
1 forceps (125mm, straight, fine point), 1 scalpel (blade length 38mm), 1 forceps (125mm, straight, blunt tip), 1 scissors (125mm straight, fine point), 2 needles in metal handle | |
10 Ea. |
Product Content Correction
Product Title
Dissecting Set in Plastic Wallet > Material: Stainless Steel