Learn More
Usbeck Laborgeraete™ Dissecting Needle
Brand:  Usbeck Laborgeraete™ 5440
Product Code. 10448571
  • 14.43 NOK / Each
Only null left
Estimated Shipment: 11-08-2025
Specifications
Laboratory Needle
Stainless Steel
50 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Usbeck Laborgeraete™ Dissecting Needle > Length: 50mm