Learn More
Schneider Gerd™ Dissecting Needle
Brand:  Schneider Gerd™ 662
Product Code. 10614385
  • 49.00 NOK / Each
Only null left
Estimated Shipment: 25-07-2025
Specifications
Dissecting
Lancet
150 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Schneider Gerd™ Dissecting Needle > Size: 150mm