Learn More
Karl Hecht™ Disposable Volume End-to-End Capillaries
Filling and discharging can be effected with or without a capillary holder.
Brand: Karl Hecht™ 40551015
This item is not returnable.
View return policy
Description
- Calibrated to contain from end to end
- Conformity certified, CE, DIN ISO 7550
- With capillary holder no. 40553001 (short tube with rubber teat)
- Accuracy of ≤ ±0.5%
- Precision of ≤ 1%
- 250 pieces per pack, 4 packs per carton
Specifications
Pipetting | |
250 per pack; 4 packs per carton | |
Single-use |
Capillary holder no. 40553001 | |
10 μL |