Learn More
Thermo Scientific™ Disposable IR Cards, 5x10cm, NaCl window, 15mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Brand: Thermo Scientific™ 043930.KJ
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Size: 5 x 10cm
- NaCl window
- 15mm aperture
Specifications
Disposable IR Cards | |
5 Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
E28U021 | Certificate of Analysis | 08/12/2021 | 043929.KM,043930.KJ |