Learn More
BRAND™ Bottletop Dispenser Thread Adapters
Constructed of Stainless Steel, PTFE, ETFE, or polypropylene.Bottletop Dispenser Thread Adapters are available in a variety of thread sizes. For use with Dispensette™, seripettor™, Titrette™, or QuikSip™ bottletop dispensers.
Brand:  BRAND™ 704270
836.46 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10518781
  • 1255.00 NOK / Each
Only null left
Estimated Shipment: 09-07-2025
Description
  • Constructed of Stainless Steel, PTFE, ETFE, or polypropylene.
  • For use with Dispensette™, seripettor™, Titrette™, or QuikSip™ bottletop dispensers.
  • Available in a variety of thread sizes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dispenser Thread Adapter
Dispensette™III, Dispensette™ Organic Remote Dispensing Unit
2 in. (outer thread), 3/4 in. (inner thread)
Stainless Steel
1 Ea.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BRAND™ Bottletop Dispenser Thread Adapters > Thread adapter; Outer diameter: 2 in.