Learn More
BRAND™ Bottletop Dispenser Thread Adapters
Constructed of Stainless Steel, PTFE, ETFE, or polypropylene.Bottletop Dispenser Thread Adapters are available in a variety of thread sizes. For use with Dispensette™, seripettor™, Titrette™, or QuikSip™ bottletop dispensers.
Brand: BRAND™ 704282
572.07 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Constructed of Stainless Steel, PTFE, ETFE, or polypropylene.
- For use with Dispensette™, seripettor™, Titrette™, or QuikSip™ bottletop dispensers.
- Available in a variety of thread sizes.
Specifications
Dispenser Thread Adapter | |
Connecting remote dispensing system with drums with GL outer thread | |
1 Ea. |
PTFE | |
Dispensette™III, Dispensette™ Organic Remote Dispensing Unit | |
32 GL (outer thread) |
Promotions
-
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-06-30
Days
21Hours
11Minutes
25Seconds
17New Offer Save Up to 78%