Learn More
BRAND™ Bottletop Dispenser Thread Adapters
Constructed of Stainless Steel, PTFE, ETFE, or polypropylene.Bottletop Dispenser Thread Adapters are available in a variety of thread sizes. For use with Dispensette™, seripettor™, Titrette™, or QuikSip™ bottletop dispensers.
Brand:  BRAND™ 704399
235.73 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10136720
  • 352.00 NOK / Each
The Ultimate Lab Appliances Promotion
Use promo code 24075 at checkout
View Available Promotions
Only null left
Estimated Shipment: 07-07-2025
Description
  • Constructed of Stainless Steel, PTFE, ETFE, or polypropylene.
  • For use with Dispensette™, seripettor™, Titrette™, or QuikSip™ bottletop dispensers.
  • Available in a variety of thread sizes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dispenser Thread Adapter
Dispensette™, seripettor™, Titrette™, QuikSip™
GL 45/38
ETFE
1 Ea.
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.
    Promo Code: 24075
    Expires2025-06-30
Product Content Correction
Product Title
BRAND™ Bottletop Dispenser Thread Adapters > Thread adapter; ETFE; 45mm/38mm