missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disodium Hydrogen Phosphate Dodecahydrate, Pharmpur™, Scharlab
Click to view available options
Quantity:
500 g
Description
- Synonyms: Sodium monohydrogen phosphate, Sodium phosphate dibasic
- Molecular Formula: Na2HPO4 • 12H20
- Molar Mass: 358.14g/mol
- CAS: 10039-32-4
- EIN ECS No.: 231-448-7
- Sodium dihydrogenphosphate (NaH2PO4): Max. 2.5%
- Reducing Substances: Passes test
Specifications
Melting Point | 35°C |
CAS | 10039-32-4 |
Quantity | 500 g |
Molecular Formula | H25Na2O16P |
Synonym | disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12 |
InChI Key | DGLRDKLJZLEJCY-UHFFFAOYSA-L |
SMILES | O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+] |
IUPAC Name | disodium;hydrogen phosphate;dodecahydrate |
Molecular Weight (g/mol) | 358.137 |
PubChem CID | 61456 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'