missing translation for 'onlineSavingsMsg'
Learn More
Disodium Hydrogen Phosphate Dihydrate, Reagent Grade, Scharlab Product Code.: 11338594

Disodium Hydrogen Phosphate Dihydrate, Reagent Grade, Scharlab

Product Code. 11338594
1 kg
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy

Product Code. 11338594

Brand: Scharlab SO03391000

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Used in analytical chemistry in phosphate buffer solutions.

  • Synonyms: Sodium monohydrogen phosphate, Sodium phosphate dibasic
  • Molecular Formula: Na2HPO4 • 2H20
  • Molar Mass: 177.99g/mol
  • CAS: 10028-24-7
  • EIN ECS No.: 231-448-7
  • Sodium dihydrogenphosphate (NaH2PO4): Max. 1.7%
  • Reducing Substances: Passes test

Specifications

Melting Point 92.5°C (release of crystalline water)
CAS 10028-24-7
pH 9.0–9.2 (5%, water)
Quantity 1 kg
Molecular Formula H5Na2O6P
MDL Number MFCD00149182
Synonym disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O
IUPAC Name disodium dihydrate hydrogen phosphate
Molecular Weight (g/mol) 177.99
PubChem CID 6451167
Formula Weight 177.99g/mol
Percent Purity 99.5% min (acidimetric)
Grade Reagent
Identification Passes Test
Loss on Drying 19.5–21.0% max (130°)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Disodium Hydrogen Phosphate Dihydrate, Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.