Learn More
Thermo Scientific™ Dionex™ PEEK Sleeves
Thermo Scientific™ Dionex PEEK Sleeves are precision cut and polished.
Brand: Thermo Scientific™ 161498
1075.12 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ Dionex PEEK Sleeves are precision cut and polished.
Specifications
For microtight union; 280 um OD | |
nanoLC | |
280 μm |
Connections with Microtight Union | |
Tubing Sleeve |