Learn More
Thermo Scientific™ Dionex™ IonPac™ NS2 5μm Analytical and Guard Column
For separation of charged analytes using mobile phase ion chromatography (MPIC) with suppressed conductivity detection.
Brand:  Thermo Scientific™ 088787
Product Code. 15560244
  • 11839.22 NOK / Each
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Description
MPIC is a method in which an ion-pairing agent in the eluent enhances retention for charged analytes on a hydrophobic column, and then is removed by the suppressor to provide selective, sensitive detection of the analytes.
  • For analysis of cations, heptafluorobutyric acid is one such agent that is amenable to suppression.
  • Excellent resolution
  • Good peak shape
  • High efficiency
  • Rugged packed bed
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4 mm
5 μm
Polar-embedded C18
150 mm
120 Å