Learn More
DIO2 Mouse anti-Human, Unconjugated, Clone: 1F2A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66813-1-IG-20UL
This item is not returnable.
View return policy
Description
The DIO2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DIO2. This antibody recognizes human antigen. The DIO2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
DIO2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
1F2A1 | |
Unconjugated | |
20 μL | |
1734 | |
IgG2a |