missing translation for 'onlineSavingsMsg'
Learn More
Dimethyl perfluoro-1,10-decanedicarboxylate, 95%, Thermo Scientific™ Product Code.: 15439049

Dimethyl perfluoro-1,10-decanedicarboxylate, 95%, Thermo Scientific™

Product Code. 15439049
5 g
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy

Product Code. 15439049

Brand: Thermo Scientific Alfa Aesar L16753.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 84750-88-9
Melting Point 33°C to 40°C
Molecular Formula C14H6F20O4
Linear Formula CH3O2C(CF2)10CO2CH3
MDL Number MFCD00236606
Quantity 5 g
Synonym dimethyl perfluoro-1,10-decanedicarboxylate, dimethyl perfluorodecane-1,10-dicarboxylate, 1,12-dimethyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluorododecanedioate, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-eicosafluorododecanedioic acid dimethyl ester, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluorododecanedioic acid dimethyl ester, dimethyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosakis fluoranyl dodecanedioate
InChI Key GHVIUCXUOSDTBP-UHFFFAOYSA-N
SMILES COC(=O)C(C(C(C(C(C(C(C(C(C(C(=O)OC)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
IUPAC Name dimethyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluorododecanedioate
Molecular Weight (g/mol) 618.166
PubChem CID 2737119
Formula Weight 618.17
Percent Purity 95%
Chemical Name or Material Dimethyl perfluoro-1,10-decanedicarboxylate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyl perfluoro-1,10-decanedicarboxylate, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.