Learn More
Dihydrotanshinone I, MedChemExpress
MedChemExpress Dihydrotanshinone I is a natural compound extracted from Salvia miltiorrhiza Bunge which has been widely used for treating cardiovascular diseases. Dihydrotanshinone I exhibits entry-blocking effect for MERS-CoV.
Brand: Medchem Express HY-N0360
This item is not returnable.
View return policy
Description
Dihydrotanshinone I is a natural compound extracted from Salvia miltiorrhiza Bunge which has been widely used for treating cardiovascular diseases. Dihydrotanshinone I exhibits entry-blocking effect for MERS-CoV.Specifications
Dihydrotanshinone I | |
Red | |
C18H14O3 | |
DMSO : 2 mg/mL (7.19 mM; Need ultrasonic) | |
278.3 | |
98.74% | |
Powder |
87205-99-0 | |
Research | |
25 mg | |
O=C(C1=C2C=CC3=C1C=CC=C3C)C(C4=C2OC[C@@H]4C)=O | |
278.3 | |
Research | |
COVID-19-anti-virus |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.