Learn More
BD Difco™ Disposable Inoculating Needles and Loops
Sterile, disposable inoculating needles do not require flaming
Brand: BD 220214
Includes: Evans Blue Dye Certificate of Calibration with every box of 1,000 loops, confirming their accuracy and consistency
Description
- Made from a unique plastic resin for controlled flexibility
- Resist cutting the agar surface, yet provide sufficient firmness to pick stubborn colonies
- Free of lubricants, oils and electrostatic charges
- Color coded
Specifications
Single-use | |
Plastic Resin | |
Light Green | |
Evans blue dye certificate of calibration |
1 μL | |
Sterile | |
Inoculating Loop | |
250/Pk. |