Learn More
BD Difco™ Antibiotic Medium
BD Difco™ Antibiotic Medium No. 12 is a microbiological assay of antibiotics.
Brand: BD 266910
This item is not returnable.
View return policy
Description
- Antibiotic assay media are used for determining antibiotic potency by the microbiological assay technique.
- Antibiotic medium 12 is prepared with the grove and randall formula. It is used for the assay of amphotericin B.
- Recommended for use in preparing test plates for the cylinder plate assay of nystatin and anisomycin.
- Certifications: Antibiotic media are prepared according to the specifications of the USP, European Pharmacopeia, and AOAC International.
Specifications
Dehydrated | |
500 g |