Learn More
Scharlab DIETYLAMIN PA 1L
Brand:  Scharlab DI04861000
DIETYLAMIN PA 1L
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11354014
  • 912.00 NOK / 1 L
Product Title
Scharlab DIETYLAMIN PA 1L >