missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylenetriaminepentaacetic acid, 98+%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
5kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent

Specifications
Specifications
Melting Point | 220.0°C |
Color | White |
Density | 1.5600g/mL |
Flash Point | 200°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98+% |
Packaging | Plastic bottle |
Quantity | 250 g |
Beilstein | 04,III,119 |
Merck Index | 15,7237 |
Show More |
Safety and Handling


Product Identifier
- Diethylenetriaminepentaacetic acid
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360D-May damage the unborn child.
- H373-May cause damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Diethylenetriaminepentaacetic acid, 98+% >
Spot an opportunity for improvement?Share a Content Correction