missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Malonate, MP Biomedicals
Click to view available options
Quantity:
100 g
Specifications
CAS | 105-53-3 |
Molecular Formula | C7H12O4 |
Quantity | 100 g |
Synonym | diethyl malonate, malonic ester, propanedioic acid, diethyl ester, dicarbethoxymethane, malonic acid, diethyl ester, carbethoxyacetic ester, malonic acid diethyl ester, 1,3-diethyl propanedioate, ethyl propanedioate, ethyl malonate van |
InChI Key | IYXGSMUGOJNHAZ-UHFFFAOYSA-N |
SMILES | CCOC(=O)CC(=O)OCC |
IUPAC Name | diethyl propanedioate |
Molecular Weight (g/mol) | 160.169 |
PubChem CID | 7761 |
Formula Weight | 160.17 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'