missing translation for 'onlineSavingsMsg'
Learn More

Diethyl bromomalonate, 90+%, Thermo Scientific Chemicals

422.00 NOK - 2555.00 NOK

Chemical Identifiers

CAS 685-87-0
Molecular Formula C7H11BrO4
Molecular Weight (g/mol) 239.065
MDL Number MFCD00009138
InChI Key FNJVDWXUKLTFFL-UHFFFAOYSA-N
Synonym diethyl bromomalonate, diethyl 2-bromomalonate, bromomalonic acid diethyl ester, ethyl bromomalonate, diethylbromomalonate, propanedioic acid, bromo-, diethyl ester, 1,3-diethyl 2-bromopropanedioate, diethyl bromopropanedioate, malonic acid, bromo-, diethyl ester, propanedioic acid, 2-bromo-, 1,3-diethyl esterShow MoreShow Less
PubChem CID 69637
IUPAC Name diethyl 2-bromopropanedioate
SMILES CCOC(=O)C(C(=O)OCC)Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11304509
SDS View Documents
Thermo Scientific Alfa Aesar
A10965.14
25 g
422.00 NOK
25g
Please to purchase this item. Need a web account? Register with us today!
11494393
SDS View Documents
Thermo Scientific Alfa Aesar
A10965.18
50 g
714.00 NOK
50g
Please to purchase this item. Need a web account? Register with us today!
11404403
SDS View Documents
Thermo Scientific Alfa Aesar
A10965.30
250 g
2555.00 NOK
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Diethyl bromomalonate is used in perfumes. It is also used to synthesize other compounds such as barbiturates, artificial flavorings, vitamin B1, and vitamin B6.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl bromomalonate is used in perfumes. It is also used to synthesize other compounds such as barbiturates, artificial flavorings, vitamin B1, and vitamin B6.

Solubility
Does not mix well with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store at room temperature. Incompatible with strong oxidizing agents and strong bases.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 211-683-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethyl bromomalonate, 90+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.