Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
U16J006 | Certificate of Analysis | 06/09/2023 | 032546.A1,032546.30 |
S28J020 | Certificate of Analysis | 14/07/2023 | 032546.A1,032546.30 |
R03J031 | Certificate of Analysis | 10/05/2023 | 032546.A1 |
TB642446 | Certificate of Analysis | 10/02/2023 | 032470.M1,032470.K2,032470.0F,032546.A1,032470.K7 |
P05I794 | Certificate of Analysis | 17/01/2023 | 032470.M1,032470.K2,032470.0F,032546.A1,032470.K7 |
Product Title
Dicyclopentadiene, typically 95%, stab., Thermo Scientific™ >