Learn More
Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
Shop All Fisher Chemical ProductsDescription
CH2Cl2, CAS Number-75-09-2, aerothene mm, methane, dichloro, methane dichloride, methylene bichloride, methylene chloride, methylene dichloride, solaesthin, freon 30, narkotil, solmethine, 2.5L, 39 deg.C, CHEBI:15767, Colorless, 84.93g/mol, YMWUJEATGCHHMB-UHFFFAOYSA-N, dichloromethane, -97 deg.C

Specifications
Specifications
Melting Point | -97°C |
Color | Colorless |
Boiling Point | 39°C |
Quantity | 2.5 L |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 350 mbar at 20°C |
Viscosity | 0.43 mPaS at 20°C |
Formula Weight | 84.93 g/mol |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
2539072 | Certificate of Analysis | 24/05/2025 | D/1852/PB15X,D/1852/17,D/1852/17X,D/1852/27,D/1852/15,D/1852/17Z,D/1852/25,D/1852/24,D/1852/15Z,D/1852/PB17,D/1852/PB15,D/1852/21,D/1852/PC15 |
2538216 | Certificate of Analysis | 24/05/2025 | D/1852/PB15X,D/1852/17,D/1852/17X,D/1852/27,D/1852/15,D/1852/17Z,D/1852/25,D/1852/24,D/1852/15Z,D/1852/PB17,D/1852/PB15,D/1852/21,D/1852/PC15 |
2534176 | Certificate of Analysis | 24/05/2025 | D/1852/PB15X,D/1852/17,D/1852/17X,D/1852/27,D/1852/15,D/1852/17Z,D/1852/25,D/1852/24,D/1852/15Z,D/1852/PB17,D/1852/PB15,D/1852/21,D/1852/PC15 |
2532972 | Certificate of Analysis | 24/05/2025 | D/1852/PB15X,D/1852/17,D/1852/17X,D/1852/27,D/1852/15,D/1852/17Z,D/1852/25,D/1852/24,D/1852/15Z,D/1852/PB17,D/1852/PB15,D/1852/21,D/1852/PC15 |
2532769 | Certificate of Analysis | 24/05/2025 | D/1852/PB15X,D/1852/17,D/1852/17X,D/1852/27,D/1852/15,D/1852/17Z,D/1852/25,D/1852/24,D/1852/15Z,D/1852/PB17,D/1852/PB15,D/1852/21,D/1852/PC15 |
Safety and Handling


- Dichloromethane
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- H351-Suspected of causing cancer.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.