Learn More
Dibenzoyl peroxide, 75%, remainder water
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 104.0°C to 106.0°C |
Color | White |
Packaging | Plastic bottle |
Linear Formula | (C6H5CO)2O2 |
Quantity | 5 g |
Beilstein | 09, 179 |
Fieser | 01,196; 04,122; 05,182; 06,160; 07,89; 12,157; 15,146; 17,367 |
Merck Index | 15, 1119 |
Solubility Information | Solubility in water: insoluble. Other solubilities: sparingly soluble in alcohol,soluble in benzene,chloroform,ether and acetone,1g/40 mL carbon disulfide,1g/40 mL olive oil |
Formula Weight | 242.23 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
A0472938 | Certificate of Analysis | 22/05/2025 | 211780050,211780010,211781000 |
A0465222 | Certificate of Analysis | 17/05/2025 | 211780050,211780010,211781000 |
A0471704 | Certificate of Analysis | 17/05/2025 | 211780050,211780010,211781000 |
A0468010 | Certificate of Analysis | 31/01/2025 | 211780050,211780010,211781000 |
A0468009 | Certificate of Analysis | 31/01/2025 | 211780050,211780010,211781000 |
Safety and Handling



- Dibenzoyl peroxide, 75%, remainder water
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Organic peroxide Type C & D
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H319-Causes serious eye irritation.
- H242-Heating may cause a fire.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.