Learn More
Diamond powder, synthetic, <1 micron, 99.9% (metals basis) Product Code.: 11356718
Diamond powder, synthetic, <1 micron, 99.9% (metals basis)
Product Code. 11356718
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11356718
Brand: Thermo Scientific Alfa Aesar 013401.06
Request Bulk or Custom Format
Diamond powder is generally used as an abrasive to grind and polish hard surfaces
Diamond powder is generally used as an abrasive to grind and polish hard surfaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Diamond powder
Name Note Synthetic
Physical Form Powder
Quantity 5 g
Assay Percent Notes (metals basis)
MDL Number MFCD00211867
Percent Purity 99.9%
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberF14W038 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code013401.06,013401.03
Lot NumberE22R025 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code013401.06,013401.03
Lot NumberG13X055 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code013401.06,013401.03
Lot NumberL03Z022 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code013401.06,013401.03
Lot NumberG02P12 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code013401.06,013401.03
Product Title
Diamond powder, synthetic, <1 micron, 99.9% (metals basis) >