missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diamide, MP Biomedicals
Click to view available options
Quantity:
100 mg
500 mg
Description
- Diamide is used as a thiol oxidizing agent
- It has also been used to titrate protein glutathiolation to discriminate from other oxidative protein modifications
- Diamide treatment increased protein glutathiolation in a concentration-dependent manner and had comparably little effect on protein-protein disulfide formation
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 104°C to 115°C |
Color | Yellow |
Linear Formula | C6H12N4O2 |
Quantity | 500 mg |
Solubility Information | Soluble in water. |
Formula Weight | 172.19 |
Physical Form | Crystalline |
Chemical Name or Material | DIAMIDE |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'