missing translation for 'onlineSavingsMsg'
Learn More
diABZI STING agonist-1 trihydrochloride, MedChemExpress Product Code.: 16497447

diABZI STING agonist-1 trihydrochloride, MedChemExpress

Product Code. 16497447
5 mg, 5mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16497447

Brand: Medchem Express HY112921B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress diABZI STING agonist-1 (trihydrochloride) is a selective stimulator of interferon genes (STING) receptor agonist, with EC50s of 130, 186 nM for human and mouse, respectively.

diABZI STING agonist-1 (trihydrochloride) is a selective stimulator of interferon genes (STING) receptor agonist, with EC50s of 130, 186 nM for human and mouse, respectively.

Specifications

Chemical Name or Material diABZI STING agonist-1 trihydrochloride
CAS 2138299-34-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C42H54Cl3N13O7
Quantity 5 mg
Solubility Information H2O : 100 mg/mL (104.24 mM; Need ultrasonic) ∣DMSO : 90 mg/mL (93.82 mM; Need ultrasonic)
SMILES O=C(N)C1=CC(N=C(NC(C2=CC(C)=NN2CC)=O)N3C/C=C/CN4C(NC(C5=CC(C)=NN5CC)=O)=NC6=C4C(OCCCN7CCOCC7)=CC(C(N)=O)=C6)=C3C(OC)=C1.[E].Cl.Cl.Cl
Molecular Weight (g/mol) 959.32
Formula Weight 959.32
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
diABZI STING agonist-1 trihydrochloride, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.