missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-Sodium Tetraborate Decahydrate, 99+%, Extra Pure, SLR, Fisher Chemical™
466.00 NOK - 1155.00 NOK
Chemical Identifiers
CAS | 1303-96-4 |
---|---|
Molecular Formula | B4Na2O7·10H2O |
MDL Number | 149193 |
Description
Chemical Identifiers
1303-96-4 | |
B4Na2O7·10H2O | |
149193 |
Specifications
75°C | |
White | |
Powder/Solid | |
149193 | |
Plastic powder jar |
1303-96-4 | |
9 | |
B4Na2O7·10H2O | |
381.36g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Sodium tetraborate decahydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
Hazard Statement
- H319-Causes serious eye irritation.
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Di-Sodium Tetraborate Decahydrate, 99+%, Extra Pure, SLR, Fisher Chemical™